Learn More
Selleck Chemical LLC Linoleic acid
SDP
Supplier:  Selleck Chemical LLC S582125MG
Encompass_Preferred
Linoleic acid (9 12-octadecadienoic acid) is an essential fatty acid used in the biosynthesis of arachidonic acid and found in the lipids of cell membranes
Catalog No. 50-000-03928