Learn More
Cayman Chemical LInoleIc AcId-13C18 100ug
SDP
Catalog No. 502989580
Encompass_Preferred
An internal standard for the quantification of linoleic acid by GC- or LC-MS
Catalog No. 50-298-9580 Supplier Cayman Chemical Supplier No. 31159100 UG
An internal standard for the quantification of linoleic acid by GC- or LC-MS