Learn More
Medchemexpress LLC Linoleic acid-13C18 | 287111-25-5 | 298.31 | 500 UG
SDP
Catalog No. 5000416210
Encompass_Preferred
Linoleic acid-13C18 is a 13C labeled version of Linoleic acid. Linoleic acid is a common polyunsaturated fatty acid (PUFA) found in plant-based oils, nuts, and seeds. It is a component of membrane phospholipids, providing structural support to maintain membrane fluidity in the transdermal water barrier of the epidermis. Linoleic acid can cause damage to red blood cells and hemoglobin through an oxidative mechanism.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of carbon have been incorporated into the molecule.
Catalog No. 50-004-16210 Supplier Medchemexpress LLC Supplier No. HYN0729S2500UG
Linoleic acid-13C18 is a 13C labeled version of Linoleic acid. Linoleic acid is a common polyunsaturated fatty acid (PUFA) found in plant-based oils, nuts, and seeds. It is a component of membrane phospholipids, providing structural support to maintain membrane fluidity in the transdermal water barrier of the epidermis. Linoleic acid can cause damage to red blood cells and hemoglobin through an oxidative mechanism.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of carbon have been incorporated into the molecule.