Learn More
Enzo Life Sciences Linoleic acid (1g). CAS: 60-33-3
SDP
Catalog No. 502010802
Encompass_Preferred
Induces adipocyte differentiation and activates PPARs in cultured cells, e.g 50 and 100μM linoleic acid, activation of PPAR-TetR fusions, U20S cells (osteosarcoma). Purity: ≥99%. Solubility: Soluble in DMSO and ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0802 Supplier Enzo Life Sciences Supplier No. BMLFA0141000
Induces adipocyte differentiation and activates PPARs in cultured cells, e.g 50 and 100μM linoleic acid, activation of PPAR-TetR fusions, U20S cells (osteosarcoma). Purity: ≥99%. Solubility: Soluble in DMSO and ethanol. Long Term Storage: -20°C.