Learn More
Cayman Chemical Linoleic Acid, 60-33-3, 50 mg
SDP
Supplier:  Cayman Chemical 9015050
Encompass_Preferred
Molecular Formula C18H32O2, Purity ≥98%, Formula Weight 280.5, Synonyms: Telfairic Acid; Linoleic acid is an essential fatty acid and one of the most abundant PUFAs in the western diet. Deficiencies in linoleic acid are linked to defective wound healing, growth retardation, and dermatitis.{2418,1017} Linoleic acid is metabolized by 15-LO in both plants and animals to form 9- and 13-HODE.{770}
Catalog No. NC9901004