Learn More
Medchemexpress LLC Linoleic acid | 60-33-3 | 280.45 | 100 MG
SDP
Catalog No. 5000416368
Encompass_Preferred
Linoleic acid is a polyunsaturated fatty acid found in plant-based oils, nuts, and seeds. It functions as a structural component of membrane phospholipids, helping to maintain membrane fluidity of the epidermal transdermal water barrier.
  • Common polyunsaturated fatty acid
  • Found in plant-based oils, nuts, and seeds
  • Part of membrane phospholipids
  • Maintains membrane fluidity of transdermal water barrier
Catalog No. 50-004-16368 Supplier Medchemexpress LLC Supplier No. HYN0729100MG
Linoleic acid is a polyunsaturated fatty acid found in plant-based oils, nuts, and seeds. It functions as a structural component of membrane phospholipids, helping to maintain membrane fluidity of the epidermal transdermal water barrier.
  • Common polyunsaturated fatty acid
  • Found in plant-based oils, nuts, and seeds
  • Part of membrane phospholipids
  • Maintains membrane fluidity of transdermal water barrier