Learn More
Medchemexpress LLC Linoleic acid | 60-33-3 | 280.45 | 50 MG
SDP
Catalog No. 5000415808
Encompass_Preferred
Linoleic acid is a common polyunsaturated fatty acid (PUFA) found in plant-based oils, nuts, and seeds. It is a component of membrane phospholipids, providing a structural function to maintain the membrane fluidity of the epidermis's transdermal water barrier. Linoleic acid can induce red blood cell and hemoglobin damage through an oxidative mechanism.
  • Common polyunsaturated fatty acid (PUFA).
  • Found in plant-based oils, nuts, and seeds.
  • Part of membrane phospholipids.
  • Functions as a structural component to maintain membrane fluidity of the epidermal transdermal water barrier.
Catalog No. 50-004-15808 Supplier Medchemexpress LLC Supplier No. HYN0729R50MG
Linoleic acid is a common polyunsaturated fatty acid (PUFA) found in plant-based oils, nuts, and seeds. It is a component of membrane phospholipids, providing a structural function to maintain the membrane fluidity of the epidermis's transdermal water barrier. Linoleic acid can induce red blood cell and hemoglobin damage through an oxidative mechanism.
  • Common polyunsaturated fatty acid (PUFA).
  • Found in plant-based oils, nuts, and seeds.
  • Part of membrane phospholipids.
  • Functions as a structural component to maintain membrane fluidity of the epidermal transdermal water barrier.