Learn More
Medchemexpress LLC Linoleic acid | 60-33-3 | MFCD00004435 | 280.4 g/mol | 10 G
SDP
Catalog No. 5000416695
Encompass_Preferred
Linoleic acid is a common polyunsaturated fatty acid (PUFA) found in plant-based oils, nuts, and seeds. It functions as a structural element of membrane phospholipids, maintaining membrane fluidity within the transdermal water barrier of the epidermis. This compound has also been observed to induce damage to red blood cells and hemoglobin through an oxidative mechanism.
  • Common polyunsaturated fatty acid (PUFA)
  • Found in plant-based oils, nuts, and seeds
  • Component of membrane phospholipids
  • Maintains membrane fluidity in the transdermal water barrier of the epidermis
  • Relevant to cardiovascular disease, metabolic disease, and cancer research
  • Applicable in microorganism, ketones, aldehydes, and acids research
  • Applicable in disease markers and endocrine diseases research
  • Relevant for endogenous metabolite, biological pesticide, and agricultural research
Catalog No. 50-004-16695 Supplier Medchemexpress LLC Supplier No. HYN072910G
Linoleic acid is a common polyunsaturated fatty acid (PUFA) found in plant-based oils, nuts, and seeds. It functions as a structural element of membrane phospholipids, maintaining membrane fluidity within the transdermal water barrier of the epidermis. This compound has also been observed to induce damage to red blood cells and hemoglobin through an oxidative mechanism.
  • Common polyunsaturated fatty acid (PUFA)
  • Found in plant-based oils, nuts, and seeds
  • Component of membrane phospholipids
  • Maintains membrane fluidity in the transdermal water barrier of the epidermis
  • Relevant to cardiovascular disease, metabolic disease, and cancer research
  • Applicable in microorganism, ketones, aldehydes, and acids research
  • Applicable in disease markers and endocrine diseases research
  • Relevant for endogenous metabolite, biological pesticide, and agricultural research