Learn More
Cayman Chemical Linoleic Acid Alkyne
SDP
Catalog No. 502888519
Encompass_Preferred
An ω-alkyne derivative of linoleic acid that can be joined with azide-containing molecules using click chemistry
Catalog No. 50-288-8519 Supplier Cayman Chemical Supplier No. 10541500 UG
An ω-alkyne derivative of linoleic acid that can be joined with azide-containing molecules using click chemistry