Learn More
Cayman Chemical Linoleic Acid Alkyne
SDP
Supplier:  Cayman Chemical 105415 MG
Encompass_Preferred
An ω-alkyne derivative of linoleic acid that can be joined with azide-containing molecules using click chemistry
Catalog No. 50-288-7210