Learn More
Cayman Chemical LInoleIc AcId AmIde 10mg
SDP
Catalog No. 502982651
Encompass_Preferred
A primary fatty acid amide derived from linoleic acid that regulates Ca2+ flux by inhibiting SERCA (IC50 = 15-53 µM)
Catalog No. 50-298-2651 Supplier Cayman Chemical Supplier No. 2105510 MG
A primary fatty acid amide derived from linoleic acid that regulates Ca2+ flux by inhibiting SERCA (IC50 = 15-53 µM)