Learn More
Cayman Chemical LInoleIc AcId AmIde 25mg
SDP
Catalog No. 502983902
Encompass_Preferred
A primary fatty acid amide derived from linoleic acid that regulates Ca2+ flux by inhibiting SERCA (IC50 = 15-53 µM)
Catalog No. 50-298-3902 Supplier Cayman Chemical Supplier No. 2105525 MG
A primary fatty acid amide derived from linoleic acid that regulates Ca2+ flux by inhibiting SERCA (IC50 = 15-53 µM)