Learn More
Cayman Chemical LInoleIc AcId-d4 500ug
SDP
Catalog No. 502915669
Encompass_Preferred
An internal standard for the quantification of linoleic acid by GC- or LC-MS
Catalog No. 50-291-5669 Supplier Cayman Chemical Supplier No. 390150500 UG
An internal standard for the quantification of linoleic acid by GC- or LC-MS