Learn More
Cayman Chemical Linoleic Acid ethyl ester
SDP
Supplier:  Cayman Chemical 10008198100 MG
Encompass_Preferred
An esterified form of linoleic acid; active against S. mutans, A. actinomycetemcomitans, P. gingivalis, S. gordonii, and S. sanguis, and C. albicans at 25 UG/ml; prevents arteriosclerosis in rabbits fed a high-cholesterol diet at 3 mg/kg
Catalog No. 50-288-7594