Learn More
Cayman Chemical Linoleic Acid ethyl ester, Purity: ≥98%, CAS: 544-35-4, 50 mg
SDP
Catalog No. 502887595
Encompass_Preferred
An esterified form of linoleic acid; active against S. mutans, A. actinomycetemcomitans, P. gingivalis, S. gordonii, and S. sanguis, and C. albicans at 25 µg/ml; prevents arteriosclerosis in rabbits fed a high-cholesterol diet at 3 mg/kg
Catalog No. 50-288-7595 Supplier Cayman Chemical Supplier No. 1000819850 MG
An esterified form of linoleic acid; active against S. mutans, A. actinomycetemcomitans, P. gingivalis, S. gordonii, and S. sanguis, and C. albicans at 25 µg/ml; prevents arteriosclerosis in rabbits fed a high-cholesterol diet at 3 mg/kg