Learn More
Cayman Chemical LInoleIc AcId methyl STR 500mg
SDP
Catalog No. 502982070
Encompass_Preferred
An esterified form of linoleic acid; has been found in several types of animal fat biodiesel and biodiesel synthesized from beef tallow, soybean oil, and babassu oil blends; has been used as a substrate to measure the antioxidant activity of β-carotene against free radical-induced lipid peroxidation
Catalog No. 50-298-2070 Supplier Cayman Chemical Supplier No. 20603500 MG
An esterified form of linoleic acid; has been found in several types of animal fat biodiesel and biodiesel synthesized from beef tallow, soybean oil, and babassu oil blends; has been used as a substrate to measure the antioxidant activity of β-carotene against free radical-induced lipid peroxidation