Learn More
Cayman Chemical Linoleic Acid QuantPAK
SDP
Catalog No. 502887041
Encompass_Preferred
The linoleic acid Quant-PAK has been designed for the convenient, precise quantification of linoleic acid by GC- or LC-MS. It includes a 50 UG vial of linoleic acid-d4 and a precisely weighed vial of unlabeled linoleic acid, with the precise weight (5 mg) indicated on the vial. This unlabeled linoleic acid can be used to quantify the linoleic acid-d4 standard by constructing a standard curve of peak intensity ratios (deuterated versus unlabeled).
Catalog No. 50-288-7041 Supplier Cayman Chemical Supplier No. 100068341 EA
The linoleic acid Quant-PAK has been designed for the convenient, precise quantification of linoleic acid by GC- or LC-MS. It includes a 50 UG vial of linoleic acid-d4 and a precisely weighed vial of unlabeled linoleic acid, with the precise weight (5 mg) indicated on the vial. This unlabeled linoleic acid can be used to quantify the linoleic acid-d4 standard by constructing a standard curve of peak intensity ratios (deuterated versus unlabeled).