Learn More
Sigma Aldrich Fine Chemicals Biosciences Linoleic Acid-Water Soluble, 10 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L590010MG
Encompass
Linoleic acid-water soluble has been used as a component of fatting media for the in vitro steatosis induction of primary human hepatocytes (PHH).
Catalog No. NC1839481