Learn More
Cayman Chemical Linoleic Acidbiotin
SDP
Catalog No. 502890230
Encompass_Preferred
An affinity probe designed to allow linoleic acid to be detected in complexes with protein-binding partners such as fatty acid binding proteins
Catalog No. 50-289-0230 Supplier Cayman Chemical Supplier No. 10010623100 UG
An affinity probe designed to allow linoleic acid to be detected in complexes with protein-binding partners such as fatty acid binding proteins