Learn More
Cayman Chemical Linoleic Acid (sodium salt), Purity: ≥95%, CAS: 822-17-3, 1 g
SDP
Catalog No. 502985772
Encompass_Preferred
An essential ω-6 PUFA; the most abundant PUFA in a variety of foods; dietary sources include vegetable oils, meats, nuts, seeds, and eggs; increases migration of IEC-6 rat intestinal epithelial cells in a wound healing assay at 30 μM; rats fed a linoleate-deficient diet exhibit decreased body weight and an increased ratio of eicosatrienoate to arachidonate in liver and serum phospholipids compared with rats fed a control diet, as well as mild scaling of forepaw skin
Catalog No. 50-298-5772 Supplier Cayman Chemical Supplier No. 219091 G
An essential ω-6 PUFA; the most abundant PUFA in a variety of foods; dietary sources include vegetable oils, meats, nuts, seeds, and eggs; increases migration of IEC-6 rat intestinal epithelial cells in a wound healing assay at 30 μM; rats fed a linoleate-deficient diet exhibit decreased body weight and an increased ratio of eicosatrienoate to arachidonate in liver and serum phospholipids compared with rats fed a control diet, as well as mild scaling of forepaw skin