Learn More
Medchemexpress LLC Linolelaidic acid | 506-21-8 | MFCD00004435 | 280.45 g/mol | C18H32O2 | 100 MG
SDP
Catalog No. 5000210645
Encompass_Preferred
Linolelaidic acid is an omega-6 trans fatty acid used as a research reagent and analytical standard. It has been studied for nutritional applications, inflammatory and anti-parasitic activities, and cellular effects such as apoptosis and cell-cycle modulation.
  • Omega-6 trans fatty acid used for research and analytical applications.
  • Reported oral activity and relevance to enteral and parenteral nutrition studies.
  • Demonstrates anti-inflammatory and anti-parasitic activity in vitro and in vivo models.
  • Useful for studies of apoptosis, cell cycle, and endothelial cell responses.
  • Available in multiple pack sizes and in solution formats for analytical use.
Catalog No. 50-002-10645 Supplier Medchemexpress LLC Supplier No. HYW071746100MG
Add to Cart
Add to Cart
Linolelaidic acid is an omega-6 trans fatty acid used as a research reagent and analytical standard. It has been studied for nutritional applications, inflammatory and anti-parasitic activities, and cellular effects such as apoptosis and cell-cycle modulation.
  • Omega-6 trans fatty acid used for research and analytical applications.
  • Reported oral activity and relevance to enteral and parenteral nutrition studies.
  • Demonstrates anti-inflammatory and anti-parasitic activity in vitro and in vivo models.
  • Useful for studies of apoptosis, cell cycle, and endothelial cell responses.
  • Available in multiple pack sizes and in solution formats for analytical use.