Learn More
Cayman Chemical γ-Linolenic Acid, Purity: ≥98%, CAS: 506-26-3, 250 mg
SDP
Catalog No. 502894262
Encompass_Preferred
γ-Linolenic acid is an ω-6 fatty acid which can be elongated to arachidonic acid for endogenous eicosanoid synthesis. It is a weak LTB4 receptor antagonist, inhibiting [3H]-LTB4 binding to porcine neutrophil membranes with a Ki of 1 µM. γ-Linolenic acid produces 53% inhibition at a 1 mg/kg dose in an in vivo model of LTB4-induced bronchoconstriction.{2361}
Catalog No. 50-289-4262 Supplier Cayman Chemical Supplier No. 90220250 MG
γ-Linolenic acid is an ω-6 fatty acid which can be elongated to arachidonic acid for endogenous eicosanoid synthesis. It is a weak LTB4 receptor antagonist, inhibiting [3H]-LTB4 binding to porcine neutrophil membranes with a Ki of 1 µM. γ-Linolenic acid produces 53% inhibition at a 1 mg/kg dose in an in vivo model of LTB4-induced bronchoconstriction.{2361}