Learn More
Cayman Chemical γ-Linolenic Acid ethyl ester, Purity: ≥98%, CAS: 31450-14-3, 1 g
SDP
Catalog No. 502893684
Encompass_Preferred
An esterified version of GLA which is less water soluble but more amenable for the formulation of GLA-containing diets
Catalog No. 50-289-3684 Supplier Cayman Chemical Supplier No. 90007381 G
An esterified version of GLA which is less water soluble but more amenable for the formulation of GLA-containing diets