Learn More
Cayman Chemical γ-Linolenic Acid ethyl ester, Purity: ≥98%, CAS: 31450-14-3, 50 mg
SDP
Catalog No. 502895321
Encompass_Preferred
An esterified version of GLA which is less water soluble but more amenable for the formulation of GLA-containing diets
Catalog No. 50-289-5321 Supplier Cayman Chemical Supplier No. 900073850 MG
An esterified version of GLA which is less water soluble but more amenable for the formulation of GLA-containing diets