Learn More
Cayman Chemical Linolenic ACD ethyl esTR 100mg
SDP
Catalog No. 502895283
Encompass_Preferred
An esterified version of GLA which is less water soluble but more amenable for the formulation of GLA-containing diets
Catalog No. 50-289-5283 Supplier Cayman Chemical Supplier No. 9000738100 MG
An esterified version of GLA which is less water soluble but more amenable for the formulation of GLA-containing diets