Learn More
Medchemexpress LLC α-Linolenic acid-13C18 | 287111-28-8 | 296.30 | 1 MG
SDP
Catalog No. 5000416693
Encompass_Preferred
α-Linolenic acid-13C18 is a 13C labeled α-Linolenic acid. This essential fatty acid, isolated from seed oils, cannot be synthesized by humans. It influences thrombosis through PI3K/Akt signaling modulation and exhibits anti-arrhythmic properties pertinent to cardiovascular disease and cancer.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Related to cardiovascular disease and cancer.
  • Modulates PI3K/Akt signaling.
  • Possesses anti-arrhythmic properties.
Catalog No. 50-004-16693 Supplier Medchemexpress LLC Supplier No. HYN0728S31MG
Add to Cart
Add to Cart
α-Linolenic acid-13C18 is a 13C labeled α-Linolenic acid. This essential fatty acid, isolated from seed oils, cannot be synthesized by humans. It influences thrombosis through PI3K/Akt signaling modulation and exhibits anti-arrhythmic properties pertinent to cardiovascular disease and cancer.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Related to cardiovascular disease and cancer.
  • Modulates PI3K/Akt signaling.
  • Possesses anti-arrhythmic properties.