Learn More
Medchemexpress LLC α-Linolenic acid-13C18 | 287111-28-8 | 296.30 | 5 MG
SDP
Catalog No. 5000417164
Encompass_Preferred
α-Linolenic acid-13C18 is the 13C labeled α-Linolenic acid. α-Linolenic acid, isolated from seed oils, is an essential fatty acid that cannot be synthesized by humans. α-Linolenic acid can affect the process of thrombotic through the modulation of PI3K/Akt signaling. α-Linolenic acid possesses anti-arrhythmic properties and is related to cardiovascular disease and cancer.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-17164 Supplier Medchemexpress LLC Supplier No. HYN0728S35MG
α-Linolenic acid-13C18 is the 13C labeled α-Linolenic acid. α-Linolenic acid, isolated from seed oils, is an essential fatty acid that cannot be synthesized by humans. α-Linolenic acid can affect the process of thrombotic through the modulation of PI3K/Akt signaling. α-Linolenic acid possesses anti-arrhythmic properties and is related to cardiovascular disease and cancer.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.