Learn More
Medchemexpress LLC α-linolenic acid-13C18 | 287111-28-8 | 99.5% | 296.30 | 13C18H30O2 | 10 MG
SDP
Catalog No. 5000296017
Encompass_Preferred
α-Linolenic acid-13C18 is a 13C18-labeled form of the essential fatty acid alpha-linolenic acid, supplied as a stable-isotope reference standard for analytical, tracer, and metabolomics studies. It is provided at high isotopic enrichment and chemical purity for use in mass spectrometry and quantitative assays.
  • Stable-isotope labeled (13C18) reference standard for tracing and quantitation.
  • High chemical purity (99.5%) for reliable analytical results.
  • Supplied as a colorless to light yellow liquid; density approx. 0.925 g/cm3.
  • Recommended storage under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Small pack sizes (for example, 10 mg) suitable for analytical workflows.
Catalog No. 50-002-96017 Supplier Medchemexpress LLC Supplier No. HYN0728S310MG
α-Linolenic acid-13C18 is a 13C18-labeled form of the essential fatty acid alpha-linolenic acid, supplied as a stable-isotope reference standard for analytical, tracer, and metabolomics studies. It is provided at high isotopic enrichment and chemical purity for use in mass spectrometry and quantitative assays.
  • Stable-isotope labeled (13C18) reference standard for tracing and quantitation.
  • High chemical purity (99.5%) for reliable analytical results.
  • Supplied as a colorless to light yellow liquid; density approx. 0.925 g/cm3.
  • Recommended storage under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Small pack sizes (for example, 10 mg) suitable for analytical workflows.