Learn More
Enzo Life Sciences Linolenic acid (18:3, n-3) (1g). CAS: 463-40-1
SDP
Supplier:  Enzo Life Sciences BMLFA0051000
Encompass_Preferred
Useful for inducing adipocyte differentiation and/or activating PPARs in cultured cells, e.g. 200 and 250μM linolenic acid for, respectively, stimulation of FA transporter transcription in FAAR (NUC I) transfected 3T3-C2 cells and differentiation of Ob1771 preadipocytes. The product is not sterile. Alternative name: Octadeca-9Z,12Z,15Z-trienoic acid. Purity: ≥99% (GC). Solubility: Soluble in ethanol, hexane and methylene chloride. Long Term Storage: -20°C.
Catalog No. 50-201-0797