Learn More
Medchemexpress LLC Α-Linolenic acid | 463-40-1 | 278.43 | 25 MG
SDP
Catalog No. 5000389407
Encompass_Preferred
α-Linolenic acid is an essential fatty acid isolated from *Perilla frutescens* that cannot be synthesized by humans. It influences thrombotic processes by modulating PI3K/Akt signaling. This compound exhibits anti-arrhythmic properties and is associated with cardiovascular disease and cancer, and converts into longer chain fatty acids eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA).
  • Essential fatty acid that cannot be synthesized by humans
  • Modulates PI3K/Akt signaling
  • Possesses anti-arrhythmic properties
  • Associated with cardiovascular disease and cancer research
  • Converts into eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA)
  • Inhibits collagen- and adrenaline-induced thrombosis
  • Suppresses A-V thrombus formation
  • Inhibits collagen stimulated platelet aggregation
Catalog No. 50-003-89407 Supplier Medchemexpress LLC Supplier No. 25MGHYN0728R
α-Linolenic acid is an essential fatty acid isolated from *Perilla frutescens* that cannot be synthesized by humans. It influences thrombotic processes by modulating PI3K/Akt signaling. This compound exhibits anti-arrhythmic properties and is associated with cardiovascular disease and cancer, and converts into longer chain fatty acids eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA).
  • Essential fatty acid that cannot be synthesized by humans
  • Modulates PI3K/Akt signaling
  • Possesses anti-arrhythmic properties
  • Associated with cardiovascular disease and cancer research
  • Converts into eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA)
  • Inhibits collagen- and adrenaline-induced thrombosis
  • Suppresses A-V thrombus formation
  • Inhibits collagen stimulated platelet aggregation