Learn More
Medchemexpress LLC α-Linolenic acid | 463-40-1 | MFCD00065720 | 278.43 | 250 MG
SDP
Catalog No. 5000389459
Encompass_Preferred
α-Linolenic acid, isolated from *Perilla frutescens*, is an essential fatty acid that humans cannot synthesize. It can influence the process of thrombosis by modulating PI3K/Akt signaling and possesses anti-arrhythmic properties. It is related to research concerning cardiovascular disease and cancer, and converts into longer chain fatty acids like eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA).
  • Essential fatty acid that humans cannot synthesize
  • Modulates PI3K/Akt signaling to impact thrombotic processes
  • Possesses anti-arrhythmic characteristics
  • Relevant to research in cardiovascular disease and cancer
  • Converts into eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA)
Catalog No. 50-003-89459 Supplier Medchemexpress LLC Supplier No. 250MGHYN0728R
α-Linolenic acid, isolated from *Perilla frutescens*, is an essential fatty acid that humans cannot synthesize. It can influence the process of thrombosis by modulating PI3K/Akt signaling and possesses anti-arrhythmic properties. It is related to research concerning cardiovascular disease and cancer, and converts into longer chain fatty acids like eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA).
  • Essential fatty acid that humans cannot synthesize
  • Modulates PI3K/Akt signaling to impact thrombotic processes
  • Possesses anti-arrhythmic characteristics
  • Relevant to research in cardiovascular disease and cancer
  • Converts into eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA)