Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC γ-Linolenic acid ethyl ester | 31450-14-3 | 99.1% | 306.48 | 5 G
SDP

Catalog No. 5000346874
Encompass_Preferred

γ-Linolenic acid ethyl ester, also known as Ethyl γ-linolenate, functions as a leukotriene B4 receptor 4 (LTB4) antagonist. It is intended for research use only.

  • Antagonist of leukotriene B4 receptor 4 (LTB4)
  • Liquid appearance
  • Colorless to light yellow
  • Molecular weight: 306.48
  • Purity: 99.14%
  • Storage conditions at 4°C, protected from light and under nitrogen
  • Soluble in DMSO at concentrations of 250 mg/mL or higher

Catalog No. 50-003-46874 Supplier Medchemexpress LLC Supplier No. HY108396100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

γ-Linolenic acid ethyl ester, also known as Ethyl γ-linolenate, functions as a leukotriene B4 receptor 4 (LTB4) antagonist. It is intended for research use only.

  • Antagonist of leukotriene B4 receptor 4 (LTB4)
  • Liquid appearance
  • Colorless to light yellow
  • Molecular weight: 306.48
  • Purity: 99.14%
  • Storage conditions at 4°C, protected from light and under nitrogen
  • Soluble in DMSO at concentrations of 250 mg/mL or higher

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.