Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linoleoyl Chloride 97.0+%, TCI America™
SDP

Catalog No. L01131G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L01131G Supplier TCI America Supplier No. L01131G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7459-33-8
Molecular Formula C18H31ClO
Molecular Weight (g/mol) 298.90
MDL Number MFCD00151403
InChI Key FBWMYSQUTZRHAT-AVQMFFATSA-N
Synonym linoleoyl chloride, linoleic acid chloride, 9z,12z-octadeca-9,12-dienoyl chloride, lineoleoyl chloride, 9,12-octadecadienoyl chloride, z,z, linolic acid chloride, linoleoyl chloride, liquid, z,z-9,12-octadecadienoyl chloride, 9z,12z octadeca-9,12-dienoyl chloride, 9,12-octadecadienoylchloride, 9z,12z
PubChem CID 9817754
IUPAC Name (9E,12E)-octadeca-9,12-dienoyl chloride
SMILES CCCCC\C=C\C\C=C\CCCCCCCC(Cl)=O

Specifications

Color Yellow
Boiling Point 168°C
Quantity 1 g
UN Number 3265
Formula Weight 298.90
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Linoleoyl Chloride
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.