Learn More
Cayman Chemical Linoleoyl Ethanolamide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1451194-69-6, 2.5 mg
SDP
Catalog No. 502894801
Encompass_Preferred
An internal standard for the quantification of linoleoyl ethanolamide by GC- or LC-MS
Catalog No. 50-289-4801 Supplier Cayman Chemical Supplier No. 90005532.5 MG
An internal standard for the quantification of linoleoyl ethanolamide by GC- or LC-MS