Learn More
Cayman Chemical LInoleoyl EthnolamIde-d4 500UG
SDP
Catalog No. 502896097
Encompass_Preferred
An internal standard for the quantification of linoleoyl ethanolamide by GC- or LC-MS
Catalog No. 50-289-6097 Supplier Cayman Chemical Supplier No. 9000553500 UG
An internal standard for the quantification of linoleoyl ethanolamide by GC- or LC-MS