Learn More
Cayman Chemical LInoleoyl GlycIn 50mg
SDP
Catalog No. 502896492
Encompass_Preferred
An endogenous homolog of arachidonyl glycine that inhibits AEA hydrolysis in FAAH-containing N18TG2 cell membranes with an IC50 value of 25 µM
Catalog No. 50-289-6492 Supplier Cayman Chemical Supplier No. 900032650 MG
An endogenous homolog of arachidonyl glycine that inhibits AEA hydrolysis in FAAH-containing N18TG2 cell membranes with an IC50 value of 25 µM