Learn More
Medchemexpress LLC Linoleyl alcohol | 506-43-4 | 266.46 | 10 MM * 1 ML
SDP
Catalog No. 5000448585
Encompass_Preferred
Linoleyl alcohol is a fatty alcohol and a structural analog of Linoleic acid, lacking an α-carboxyl group. This colorless to light yellow liquid is derived from plants and is intended for research purposes only. Studies have shown its involvement in the formation of hydroperoxyoctadecadien-1-ols and its potential in promoting weight loss and managing metabolic syndrome through esterification.
  • Structural analog of linoleic acid
  • Lacks an α-carboxyl group
  • Classified as a fatty alcohol
  • Forms 9- and 13-hydroperoxyoctadecadien-1-ols
  • Esters may promote weight loss
  • Esters may help manage metabolic syndrome
  • Colorless to light yellow liquid
  • Density of 0.8588 g/cm³
  • Derived from plants
Catalog No. 50-004-48585 Supplier Medchemexpress LLC Supplier No. HYW00562710MM/1ML
Linoleyl alcohol is a fatty alcohol and a structural analog of Linoleic acid, lacking an α-carboxyl group. This colorless to light yellow liquid is derived from plants and is intended for research purposes only. Studies have shown its involvement in the formation of hydroperoxyoctadecadien-1-ols and its potential in promoting weight loss and managing metabolic syndrome through esterification.
  • Structural analog of linoleic acid
  • Lacks an α-carboxyl group
  • Classified as a fatty alcohol
  • Forms 9- and 13-hydroperoxyoctadecadien-1-ols
  • Esters may promote weight loss
  • Esters may help manage metabolic syndrome
  • Colorless to light yellow liquid
  • Density of 0.8588 g/cm³
  • Derived from plants