Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Linopirdine

Catalog No. L13450MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L13450MG 50 mg
L13410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L13450MG Supplier Sigma Aldrich Supplier No. L13450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Stimulates release of acetylcholine and other neurotransmitters; cognitive enhancer.Sold under exclusive license from DuPont.

Specifications

CAS 105431-72-9
Molecular Formula C26H21N3O
Linear Formula C26H21N3O
Quantity 50 mg
MDL Number MFCD00867216
Synonym 1,3-Dihydro-1-phenyl-3,3-bis(4-pyridinylmethyl)-2 H-indol-2-one; DuP 996
Molecular Weight (g/mol) 391.46
Percent Purity ≥98% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.