Learn More
Medchemexpress LLC HY-W020468 5mg Medchemexpress, Linopirdine CAS:105431-72-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0204685MG
Encompass_Preferred
Medchemexpress, HY-W020468 5mg Linopirdine CAS:105431-72-9 Linopirdine (DuP 996) is an orally active, selective M-type K+ current (IM; Kv7; KCNQ Channels) inhibitor with an IC50 of 2.4 μM. Linopirdine is a TRPV1 agonist. Linopirdine, a putative cognition enhancing drug, increases acetylcholine release in rat brain tissue. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3439