Learn More
Medchemexpress LLC Linperlisib | 1702816-75-8 | MFCD31544346 | 98.9% | 588.69 g·mol⁻¹ | C28H37FN6O5S | 25 MG
SDP
Catalog No. 5000247305
Encompass_Preferred
Linperlisib is a potent, orally bioavailable, and selective inhibitor of phosphoinositide 3-kinase delta (PI3Kδ) developed for research applications. It demonstrates an IC50 of approximately 6.4 nM against PI3Kδ and is supplied as a high-purity solid suitable for in vitro and preclinical studies.
  • Potent, selective inhibitor of PI3Kδ (IC50 ≈ 6.4 nM).
  • Orally bioavailable profile useful for in vivo studies.
  • Supplied as a high-purity solid (≈98.9%).
  • Suitable for in vitro and preclinical research applications.
  • Available in multiple small-scale quantities for laboratory use.
  • Recommended storage at low temperatures to preserve stability.
Catalog No. 50-002-47305 Supplier Medchemexpress LLC Supplier No. HY10203125MG
Linperlisib is a potent, orally bioavailable, and selective inhibitor of phosphoinositide 3-kinase delta (PI3Kδ) developed for research applications. It demonstrates an IC50 of approximately 6.4 nM against PI3Kδ and is supplied as a high-purity solid suitable for in vitro and preclinical studies.
  • Potent, selective inhibitor of PI3Kδ (IC50 ≈ 6.4 nM).
  • Orally bioavailable profile useful for in vivo studies.
  • Supplied as a high-purity solid (≈98.9%).
  • Suitable for in vitro and preclinical research applications.
  • Available in multiple small-scale quantities for laboratory use.
  • Recommended storage at low temperatures to preserve stability.