Learn More
Med Vet International Medi-Vac CRD Suction Canister Liner, 1500 mL, Sealing Lid, 65651-515, Each
Catalog No. 503043065
Encompass_Preferred
CRD disposable liners are easily assembled by snapping the lid onto the liner Engineered for clinician safety, CRD lids contain integrated filters and shut-off valves The lids lock into place after assembly in order to minimize the risk of spillage during the transport process CRD disposable liners require the use of a reusable outer canister This 1500 mL suction canister liner does not include tubing
Catalog No. 50-304-3065 Supplier Med Vet International Supplier No. 243810EA
CRD disposable liners are easily assembled by snapping the lid onto the liner Engineered for clinician safety, CRD lids contain integrated filters and shut-off valves The lids lock into place after assembly in order to minimize the risk of spillage during the transport process CRD disposable liners require the use of a reusable outer canister This 1500 mL suction canister liner does not include tubing