Learn More
TARGETMOL CHEMICALS INC LINSIDOMINE HYDROCHLORID 5MG
SDP
Catalog No. 502580877
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Linsidomine hydrochloride (SIN-1 chloride) is considered as a metabolite of moxidomine with vasodilation inhibition of platelet aggregation and antiangina activity. In myocardial ischemia-reperfusion models Linsidomine hydrochloride to reduce myocardial necrosis and reperfusion induced endothelial dysfunction is associated with the cavernosal mechanism of action involved in nitric oxide release. purity: 99%
Catalog No. 50-258-0877 Supplier TARGETMOL CHEMICALS INC Supplier No. T233565MG
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Linsidomine hydrochloride (SIN-1 chloride) is considered as a metabolite of moxidomine with vasodilation inhibition of platelet aggregation and antiangina activity. In myocardial ischemia-reperfusion models Linsidomine hydrochloride to reduce myocardial necrosis and reperfusion induced endothelial dysfunction is associated with the cavernosal mechanism of action involved in nitric oxide release. purity: 99%