Learn More
Medchemexpress LLC Linsidomine hydrochloride | 16142-27-1 | 100.0% | 206.63 g/mol | C6H11ClN4O2 | 100 MG
SDP
Catalog No. 5000211203
Encompass_Preferred
Linsidomine hydrochloride is a research chemical used as a spontaneous reactive oxygen/nitrogen species (ROS/RNS) generator and peroxynitrite donor. It is applied in biochemical and cellular studies of oxidative stress, nitric oxide-related signaling, vascular function, and platelet aggregation when controlled reactive species release is required.
  • Acts as a spontaneous ROS/RNS generator and peroxynitrite donor.
  • Useful for studying oxidative stress and nitric oxide signaling pathways.
  • Exhibits vasodilatory activity and inhibits platelet aggregation in functional assays.
  • High purity solid suitable for preparation of defined stock solutions.
Catalog No. 50-002-11203 Supplier Medchemexpress LLC Supplier No. HY101200100MG
Linsidomine hydrochloride is a research chemical used as a spontaneous reactive oxygen/nitrogen species (ROS/RNS) generator and peroxynitrite donor. It is applied in biochemical and cellular studies of oxidative stress, nitric oxide-related signaling, vascular function, and platelet aggregation when controlled reactive species release is required.
  • Acts as a spontaneous ROS/RNS generator and peroxynitrite donor.
  • Useful for studying oxidative stress and nitric oxide signaling pathways.
  • Exhibits vasodilatory activity and inhibits platelet aggregation in functional assays.
  • High purity solid suitable for preparation of defined stock solutions.