Learn More
Medchemexpress LLC Linsidomine hydrochloride | 16142-27-1 | MFCD00221615 | 100.0% | 206.63 g/mol | C6H11ClN4O2 | 25 MG
SDP
Catalog No. 5000213193
Encompass_Preferred
Linsidomine hydrochloride is a research reagent that generates reactive oxygen and nitrogen species, acting as a peroxynitrite donor and nitric-oxide releaser. It is used to model oxidative and nitrative stress, and to study vasodilation, platelet aggregation, and related signaling pathways in cellular and biochemical systems.
  • Reactive nitrogen species donor and peroxynitrite generator.
  • Useful for studying oxidative and nitrative stress in cellular systems.
  • Enables examination of vasodilation and platelet aggregation mechanisms.
  • High reported purity (~99.97%) supports reproducible experimental results.
  • Solid form with recommended storage at -20°C; solutions stable at -80°C for extended storage.
Catalog No. 50-002-13193 Supplier Medchemexpress LLC Supplier No. HY10120025MG
Linsidomine hydrochloride is a research reagent that generates reactive oxygen and nitrogen species, acting as a peroxynitrite donor and nitric-oxide releaser. It is used to model oxidative and nitrative stress, and to study vasodilation, platelet aggregation, and related signaling pathways in cellular and biochemical systems.
  • Reactive nitrogen species donor and peroxynitrite generator.
  • Useful for studying oxidative and nitrative stress in cellular systems.
  • Enables examination of vasodilation and platelet aggregation mechanisms.
  • High reported purity (~99.97%) supports reproducible experimental results.
  • Solid form with recommended storage at -20°C; solutions stable at -80°C for extended storage.