Learn More
Apexbio Technology LLC Linsitinib(Synonyms: OSI-906, ASP7487, IGF-1R inhibitor OSI-906, IGF-1R/IR inhibitor OSI-906, OSI906), 10mM (in 1mL DMSO), CAS: 867160-71-2.
SDP
Catalog No. 5000060936
Encompass_Preferred
Linsitinib (CAS 867160-71-2) also known as OSI-906 is a small-molecule inhibitor targeting both insulin receptor (IR) and insulin-like growth factor-1 receptor (IGF-1R) kinases It exhibits potent inhibitory effects with IC50 values of 75 nM against IR and 35 nM against IGF-1R In vitro studies using the 3T3/hulGF-1R (LISN) cell line demonstrate that Linsitinib inhibits IGF-1R ligand-dependent autophosphorylation and downstream signaling via pERK1/2 pAKT and p-p70S6K It shows antiproliferative activity across multiple cancer cell lines and exhibits dose-dependent tumor growth inhibition in LISN xenograft animal models supporting its utility in oncology research
Catalog No. 50-000-60936 Supplier Apexbio Technology LLC Supplier No. A83345.1
Linsitinib (CAS 867160-71-2) also known as OSI-906 is a small-molecule inhibitor targeting both insulin receptor (IR) and insulin-like growth factor-1 receptor (IGF-1R) kinases It exhibits potent inhibitory effects with IC50 values of 75 nM against IR and 35 nM against IGF-1R In vitro studies using the 3T3/hulGF-1R (LISN) cell line demonstrate that Linsitinib inhibits IGF-1R ligand-dependent autophosphorylation and downstream signaling via pERK1/2 pAKT and p-p70S6K It shows antiproliferative activity across multiple cancer cell lines and exhibits dose-dependent tumor growth inhibition in LISN xenograft animal models supporting its utility in oncology research