Learn More
Selleck Chemical LLC Linsitinib (OSI-906) S1091-50mg
SDP
Catalog No. 503108402
Encompass_Preferred
Linsitinib (OSI-906) is a selective inhibitor of IGF-1R with IC50 of 35 nM in cell-free assays modestly potent to InsR with IC50 of 75 nM and no activity towards Abl ALK BTK EGFR FGFR1/2 PKA etc Phase 3
Catalog No. 50-310-8402 Supplier Selleck Chemical LLC Supplier No. S109150MG
Linsitinib (OSI-906) is a selective inhibitor of IGF-1R with IC50 of 35 nM in cell-free assays modestly potent to InsR with IC50 of 75 nM and no activity towards Abl ALK BTK EGFR FGFR1/2 PKA etc Phase 3