Learn More
Medchemexpress LLC HY-10191 100mg , Linsitinib OSI-906 CAS:867160-71-2 Purity:98%
SDP
Catalog No. 501492696
Encompass_Preferred
Medchemexpress, HY-10191 100mg Linsitinib OSI-906 CAS:867160-71-2 Formula:C26H23N5O IC50: 35 nM (IGF-1R), 75 nM (InsR) Purity:98% Linsitinib is a selective inhibitor of  IGF-1R  with  IC 50  of 35 nM, and modestly potent to InsR with IC 50 of 75 nM, and has no activity towards Abl, ALK, BTK, EGFR, FGFR1/2, PKA etc. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2696 Supplier Medchemexpress LLC Supplier No. HY10191100MG
Medchemexpress, HY-10191 100mg Linsitinib OSI-906 CAS:867160-71-2 Formula:C26H23N5O IC50: 35 nM (IGF-1R), 75 nM (InsR) Purity:98% Linsitinib is a selective inhibitor of  IGF-1R  with  IC 50  of 35 nM, and modestly potent to InsR with IC 50 of 75 nM, and has no activity towards Abl, ALK, BTK, EGFR, FGFR1/2, PKA etc. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.