Learn More
Research Products International Corp Linsmaier & Skoog Medium, Powder, Makes 5 Liters
SDP
Catalog No. 50213398
Encompass_Preferred
Basal salt mixture containing micro and macro elements with vitamins as described by Linsmaier & Skoog (1965). Package contains 4.4 grams of powder per liter of medium. Hygroscopic.
Catalog No. 50-213-398 Supplier Research Products International Corp Supplier No. L240005.0
Basal salt mixture containing micro and macro elements with vitamins as described by Linsmaier & Skoog (1965). Package contains 4.4 grams of powder per liter of medium. Hygroscopic.