Learn More
Bioworld Linsmaier & Skoog Basal Medium, w/ Sucrose & Agar, 25 L
Catalog No. 50254915
Encompass_Preferred
Linsmaier & Skoog Basal MediumContains Macronutrients, Micronutrients, and Vitamins as described by Linsmaier & Skoog (1965) with addition of Sucrose and AgarCytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 41.43 g/LpH: 4.25–5.25Partially soluble in water Medium may be sold as Murashige & Skoog (MS) Medium with Minimal Organics (MSMO) by some media suppliers
Catalog No. 50-254-915 Supplier Bioworld Supplier No. 306301683
Linsmaier & Skoog Basal MediumContains Macronutrients, Micronutrients, and Vitamins as described by Linsmaier & Skoog (1965) with addition of Sucrose and AgarCytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 41.43 g/LpH: 4.25–5.25Partially soluble in water Medium may be sold as Murashige & Skoog (MS) Medium with Minimal Organics (MSMO) by some media suppliers