Learn More
Bioworld Linsmaier & Skoog Basal Medium, w/ Sucrose, pH Adjusted and Buffered, 50 L
Catalog No. 50254922
Encompass_Preferred
Linsmaier & Skoog Basal MediumContains Macronutrients, Micronutrients, and Vitamins as described by Linsmaier & Skoog (1965) with addition of SucroseBuffered with MESCytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 35.53 g/LpH: 5.25–6.25Soluble in water Medium may be sold as Murashige & Skoog (MS) Medium with Minimal Organics (MSMO) by some media suppliers
Catalog No. 50-254-922 Supplier Bioworld Supplier No. 306301704
Linsmaier & Skoog Basal MediumContains Macronutrients, Micronutrients, and Vitamins as described by Linsmaier & Skoog (1965) with addition of SucroseBuffered with MESCytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 35.53 g/LpH: 5.25–6.25Soluble in water Medium may be sold as Murashige & Skoog (MS) Medium with Minimal Organics (MSMO) by some media suppliers